
4-Hydroxy-3-pyridinesulphonic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:51498-37-4
:
Formula:C₅H₅NO₄S
MDL Number:MFCD02093473
ADR:8,III
:175.1625 g/mol
EINECS:257-243-2
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Solubility in hot Water:very faint turbidity
Specification Test Results
Frequently Bought Together