4-Hydroxy-3-methoxybenzylamine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:7149-10-2
:
Formula:C₈H₁₂ClNO₂
MDL Number:MFCD00012864
:189.64 g/mol
Melting Pt:220°C
EINECS:230-468-3
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Frequently Bought Together