
4-Carboxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:14047-29-1
:
Formula:C₇H₇BO₄
MDL Number:MFCD00151801
:165.94 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):97.0 to 112.0 %
Specification Test Results
Frequently Bought Together