0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Dodecyl methyl sulphide
Dodecyl methyl sulphide ≥98.0% (by GC)
Dodecyl methyl sulphide ≥98.0% (by GC)
:
TCI
Dodecyl methyl sulphide ≥98.0% (by GC)
:
TCI
:
3698-89-3
:
:
C₁₃H₂₈S
:
MFCD00015073
:
216.41 g/mol
:
110°C
:
–2°C
:
223-028-7
Appearance:
Colorless to Almost colorless clear liquid
Purity (GC):
min. 98.0 %