
5-Indoleboronic acid pinacol ester ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:269410-24-4
:
Formula:C₁₄H₁₈BNO₂
MDL Number:MFCD03789263
:243.1092 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:124.0 to 128.0 deg-C