Difurfuryl sulphide 96%
Catalog # ACRO265080050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to brown Low melting solid or liquid
- Infrared spectrum:Conforms
- GC:≥95 %
Specification Test Results
About this item
:
Boiling Pt:135…143 °C (14 mmHg)
Melting Pt:30…32 °C
Flash Pt:>110 °C
:194.2500 g/mol
Storage Temperature:Ambient
EINECS:237-172-3
CAS Number:13678-67-6
Formula:C₁₀H₁₀O₂S
MDL Number:MFCD00010081