
trans-4-(Aminomethyl)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1197-18-8
:
Formula:C₈H₁₅NO₂
MDL Number:MFCD00001466
:157.21 g/mol
Merck Index:14,09569
EINECS:214-818-2
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Water:almost transparency