


Ebselen
Catalog # J63190.MA
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
Specifications
About this item
:
:274.20 g/mol
Merck Index:14,3486
Melting Pt:180…181 °C
Storage Temperature:Freezer
UN:3283
ADR:6.1,III
:GHS06
:GHS08
:GHS09
CAS Number:60940-34-3
Formula:C₁₃H₉NOSe
MDL Number:MFCD00210937