Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Methoxy-2-(trifluoromethyl)phenylboronic acid
4-Methoxy-2-(trifluoromethyl)phenylboronic acid
4-Methoxy-2-(trifluoromethyl)phenylboronic acid
Irritant
4-Methoxy-2-(trifluoromethyl)phenylboronic acid
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
4-Methoxy-2-(trifluoromethyl)phenylboronic acid
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
313546-16-6
:
Formula:
C₈H₈BF₃O₃
MDL Number:
MFCD03095346
:
219.95 g/mol
Melting Pt:
168…172 °C
:
GHS07
Related Information
Documentation
SDS
msds_ALFAH27062
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation