Order Entry
Finland
ContactUsLinkComponent
Dispensing bottle
Dispensing bottle
Catalog # 215-6000
Supplier:  KARTELL
undefined
Dispensing bottle
Catalog # 215-6000
Supplier:  KARTELL

Specifications

  • Ext.Ø:
    77 mm
  • Capacity:
    500 ml
  • Height:
    229 mm
  • Pk:
    5

Specifications

About this item

HDPE/PP.

  • Good chemical resistance
  • Good temperature resistance

Dispensing bottle suitable for liquids single and/or continuous dispensing.