Order Entry
Finland
ContactUsLinkComponent
Sleeve protector, cut resistant, Aracut® Sleeve 14
Sleeve protector, cut resistant, Aracut® Sleeve 14
Catalog # 112-2919
Supplier:  HONEYWELL SAFETY
undefined
Sleeve protector, cut resistant, Aracut® Sleeve 14
Catalog # 112-2919
Supplier:  HONEYWELL SAFETY
Specifications
  • Garment fit:
    Regular
  • Garment application:
    Mechanical protection
  • Gender:
    Unisex
  • Risk area:
    Heat,
    Cuts
  • Colour:
    Yellow
  • Length:
    350 mm
  • Fabric:
    Kevlar®
  • Pk:
    1
Specifications
About this item
This Kevlar® thread knitted sleeve with thumbhole provides protection of the forearm against cuts and punctures.
  • Knitting technique helps to obtain a cuff close to the hand, with good dexterity
  • Kevlar® fibre offers good cutting resistance and protection against thermal hazards (contact with heat)
  • Two layers of Kevlar®: Greater mechanical and thermal performance of the cuff
  • Reinforced thumb gives the cuff better support
Gauge: 10
EN 388 (132X), EN 407 (X1XXXX); Cat. II
Certifications: EN 388 (132X), EN 407 (X1XXXX); Cat. II