Nitrile gloves, Showa 707FL
: Showa
These lined nitrile gloves with rolled cuffs protect the hand from oils, hydrocarbons, grease and abrasion.
- Non sterile, powder-free, smooth finish
- Comfortable flock lining for continuous wear
- Impermeable for working in damp or greasy environments
- Tractor tread finish and 'slip-on' treatment provide good grip
- Easy to put on and remove
- Semi-disposable; can be used once or re-used
Suitable for manufacturing, laboratories, food processing, light duty maintenance, clean up tasks, paint applications, fuel handling and refineries.
: EN ISO 374-1 Type A (JKLOPT), EN ISO 374-5; Cat. III; AQL 1,5
Food contact approved.
Food contact approved.
- Catalog No:
- 112-2749
- 112-2775
- 112-2773
- 112-2776
- 112-2774
- 112-2748
- Model:
- 707FL
- 707FL
- 707FL
- 707FL
- 707FL
- 707FL
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN ISO 374-1 Type A (JKLOPT), EN ISO 374-5
- EN ISO 374-1 Type A (JKLOPT), EN ISO 374-5
- EN ISO 374-1 Type A (JKLOPT), EN ISO 374-5
- EN ISO 374-1 Type A (JKLOPT), EN ISO 374-5
- EN ISO 374-1 Type A (JKLOPT), EN ISO 374-5
- EN ISO 374-1 Type A (JKLOPT), EN ISO 374-5
- Food contact approved:
- +
- +
- +
- +
- +
- +