Earmuffs, Peltor™ Optime™ III
: 3M
These earmuffs have been developed for use in extremely high noise environments, particularly those dominated by low frequency sounds. The protection is based on a technology with a double casing, minimising resonance in the holder casing resulting in maximum high frequency attenuation.
- Broad sealing rings filled with soft plastic foam for optimum fit and low contact pressure
- Good speech intelligibility
- Easy to replace cushions and inserts help keep them hygienically clean
- Helmet-mounted version is designed to fit a wide range of industrial safety helmets
SNR: 35 dB (headband/neckband); 34 dB (helmet-mounted)
: EN 352-1:1993 (neckband version); EN 352-3:2002 (helmet-mounted version)
: *Suitable for all Peltor™ helmets, except G2000