Bowls, disposable
Supplier: Sampling Systems
Certificates
About this item
HDPE, white.
- Moulded and packed in a cleanroom
- Individually bagged
- BSE- and TSE-free
- Full batch traceability
Ideal for handling and mixing powders and liquids. With spout and grip.
Certifications: FDA acceptable CFR 177.1520, EU Regulations 10/2011 compliant