SPE columns, CHROMABOND® HR-XC
CHROMABOND® HR-XC is a benzenesulfonic acid modified polymeric cation exchanger for SPE.
- Base material: Spherical PS/DVB copolymer with a large specific surface and a high pH stability of 1 to 14
- Cation exchange capacity 1,0 meg/g
- Outstanding recovery rates especially for basic analytes
Recommended applications include separation of basic active ingredients from strongly matrix-contaminated samples like urine, plasma or serum as well as basic substances like fungicides from food. Also suited for basic analytes like amines and bases with pKa 2 to 10.
- Catalog No:
- MANA730049
- MANA730956
- MANA730969
- MANA730955
- MANA731756
- 705-1110
- MANA730969P45
- MANA730953
- MANA730952P45
- MANA730956P45
- MANA730049P45
- MANA731757
- MANA731755
- 705-1109
- pH range:
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- 1 - 14
- Surface area:
- 800 m²/g
- 800 m²/g
- 800 m²/g
- 800 m²/g
- 800 m²/g
- 800 m²/g
- 800 m²/g
- 800 m²/g
- 800 m²/g
- 800 m²/g
- 800 m²/g
- 800 m²/g
- 800 m²/g
- 800 m²/g