Order Entry
Finland
ContactUsLinkComponent
SPE columns, CHROMABOND® PSA
SPE columns, CHROMABOND® PSA
Supplier:  MACHEREY-NAGEL
Certificates
undefined
SPE columns, CHROMABOND® PSA
Supplier:  MACHEREY-NAGEL
Specifications
  • Catalog No:
  • MANA730464
  • MANA730462
  • pH range:
  • 2 - 8
  • 2 - 8
  • Surface area:
  • 500 m²/g
  • 500 m²/g
Specifications