Gel permeation chromatography columns, PLgel Olexis™
Supplier: VARIAN
Certificates
About this item
PLgel Olexis is optimized for the analysis of high molecular weight polymers at elevated temperatures, particularly polyolefins and high performance polymers.
- Optimized design for analysis of polyolefins and high performance polymers
- Fewer replacements, delays, and failed runs due to column degradation
- Operates at temperatures up to 220 °C
The column is packed with 13 µm particles designed to strike the best balance between efficiency, lifetime, and resolution without the risk of shear degradation of samples.