HPLC columns, NUCLEOSIL® SA
NUCLEOSIL® SA is a sulfonic acid modified silica phase (SCX) for HPLC.
- Robust spherical silica gel
- Cationic phase for ion exchange chromatography
- Separation mechanism based on strong ion exchange interactions
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
Capacity: ~1 meq/g
- Catalog No:
- 554-5808
- MANA720182.46
- MANA720709.46
- MANA720028.46
- MANA720097.40
- 554-5803
- Carbon load:
- 6,5%
- 6,5%
- 6,5%
- 6,5%
- 6,5%
- 6,5%
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- USP code:
- L9
- L9
- L9
- L9
- L9
- L9