
4-(2-Pyridylazo)resorcinol ≥98.0% (by HPLC, titration analysis) spectrophotometric reagent for transition metals
: TCI
:1141-59-9
:
:C₁₁H₉N₃O₂
:MFCD00006256
:215.21 g/mol
:214-528-6
:2811
:6.1,III
:GHS06
- Appearance:Orange to Amber to Dark red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Sensitiveness:Coloring by copper sulfate solution. Fading by EDTA2Na.