
Imidazole-4,5-dicarboxylic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAI0003-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
About this item
:
:156.10 g/mol
EINECS:209-327-5
:GHS07
CAS Number:570-22-9
Formula:C₅H₄N₂O₄
MDL Number:MFCD00005200