Order Entry
Finland
ContactUsLinkComponent
Acridine orange (hemizinc salt)
Health
Irritant
Acridine orange (hemizinc salt)
Catalog # TCIAA0132-25G
Supplier:  TCI
CAS Number:  
undefined
Health
Irritant
Acridine orange (hemizinc salt)
Catalog # TCIAA0132-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Dark red to Brown powder to crystal
  • Lambda max.:
    490.0 to 494.0 nm (H2O)
  • Absorbance (E1%1cm):
    min. 900 (H2O, 490.0 to 494.0 nm)

Specification Test Results

About this item

 :
 :369.96 g/mol
EINECS:233-353-6
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:10127-02-3
Formula:C₁₇H₂₀ClN₃·HCl·½ZnCl₂
MDL Number:MFCD00081043