


Dichloro(chloromethyl)methylsilane ≥97.0% (by GC)
Catalog # TCIAC1358-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
About this item
:
:163.51 g/mol
Boiling Pt:122°C
Flash Pt:35°C
Storage Temperature:Refrigerator
EINECS:216-319-5
UN:3362
ADR:6.1,II
:GHS02
:GHS05
:GHS06
CAS Number:1558-33-4
Formula:C₂H₅Cl₃Si
MDL Number:MFCD00000874