Benzoyl peroxide ≥75.0% (by titrimetric analysis) wetted with ca. 25% water (unit weight on dry weight basis)
TCIAB3152-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (Iodometric Titration):75.0 to 78.0 %
:
:242.23 g/mol
:14,01116
:5.2,
:Moistened with water
:202-327-6
:3104
:GHS02
:GHS07
:GHS09
:Refrigerator
:94-36-0
:C₁₄H₁₀O₄
:MFCD00003071