Order Entry
Finland
ContactUsLinkComponent
Ethyl acetate ≥99.5% (by GC) for spectrophotometry
Flammable
Irritant
Ethyl acetate ≥99.5% (by GC) for spectrophotometry
Catalog # TCIAA0030-250ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Ethyl acetate ≥99.5% (by GC) for spectrophotometry
Catalog # TCIAA0030-250ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.5 %
  • Absorbance (260nm):
    max. 0.300
  • Absorbance (270nm):
    max. 0.020
  • Absorbance (280nm):
    max. 0.010

Specification Test Results

About this item

 :
 :88.11 g/mol
Merck Index:14,03757
Boiling Pt:77°C
Melting Pt:–83 °C
Density:0,902 g/cm³ (20 °C)
Flash Pt:–3°C
Storage Temperature:Ambient
EINECS:205-500-4
UN:1173
ADR:3,II
REACH:01-2119475103-46
 :GHS02
 :GHS07
CAS Number:141-78-6
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171