Order Entry
Finland
ContactUsLinkComponent
1,3-Dichloro-2-butene cis- and trans-mixture ≥95.0% (by GC)
Flammable
Toxic
1,3-Dichloro-2-butene cis- and trans-mixture ≥95.0% (by GC)
Catalog # TCIAD0351-25G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Toxic
1,3-Dichloro-2-butene cis- and trans-mixture ≥95.0% (by GC)
Catalog # TCIAD0351-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 % (total of isomers)

Specification Test Results

About this item

 :
 :125 g/mol
Boiling Pt:128 °C (1013 hPa)
Melting Pt:–98 °C
Density:1,161 g/cm³ (20 °C)
Flash Pt:27°C
EINECS:213-138-3
UN:1992
ADR:3,III
 :GHS02
 :GHS06
CAS Number:926-57-8
Formula:C₄H₆Cl₂
MDL Number:MFCD00000861