Bismarck brown Y
Catalog # TCIAB0780-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Red to Dark blue to Black powder to crystal
- Lambda max.:457.0 to 463.0 nm (H2O)
- Absorbance (E1%1cm):min. 250 (H2O, 457.0 to 463.0 nm)
- Drying loss:max. 10.0 %
Specification Test Results
About this item
:
:419.32 g/mol
Merck Index:14,01253
Storage Temperature:Ambient
EINECS:233-314-3
CAS Number:10114-58-6
Formula:C₁₈H₁₈N8·2HCl
MDL Number:MFCD00012977