

ω-Bromo-3-methoxyacetophenone ≥98.0% (by titrimetric analysis)
: TCI
:5000-65-7
:
:C₉H₉BrO₂
:MFCD00000199
:229.07 g/mol
:285 °C (1013 hPa)
:63…64.3 °C
:Refrigerator
:225-666-1
:3261
:8,II
:GHS05
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:61.0 to 65.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together