

ω-Bromo-3-methoxyacetophenone ≥99.0% (by titrimetric analysis) for HPLC labeling
Catalog # TCIAA5505-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 99.0 %
- Melting point:62.0 to 64.0 deg-C
- Solubility in Methanol:almost transparency
- Effect test of HPLC derivatization:Effective as labeling agent for Linoleic Acid , Stearic Acid
Specification Test Results
About this item
:
:229.07 g/mol
Boiling Pt:285 °C (1013 hPa)
Melting Pt:63…64.3 °C
Storage Temperature:Refrigerator
EINECS:225-666-1
UN:3261
ADR:8,II
:GHS05
:GHS07
CAS Number:5000-65-7
Formula:C₉H₉BrO₂
MDL Number:MFCD00000199