Order Entry
Finland
ContactUsLinkComponent
Dipropyl sulphide ≥98.0% (by GC)
Flammable
Irritant
Dipropyl sulphide ≥98.0% (by GC)
Catalog # TCIAP0532-25ML
Supplier:  TCI
CAS Number:  
Flammable
Irritant
Dipropyl sulphide ≥98.0% (by GC)
Catalog # TCIAP0532-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :118.24 g/mol
Merck Index:14,07868
Boiling Pt:142°C
Melting Pt:–102 °C
Density:0,83 g/cm³ (20 °C)
Flash Pt:31°C
Storage Temperature:Ambient
EINECS:203-873-8
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:111-47-7
Formula:C₆H₁₄S
MDL Number:MFCD00009379