
3-Fluoro-4-(trifluoromethyl)aniline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:69411-68-3
:
Formula:C₇H₅F₄N
MDL Number:MFCD00190120
:179.12 g/mol
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:54.0 to 58.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together