4-tert-Butylphthalic anhydride 95%, Sigma-Aldrich®
Catalog # 454648-5G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to off-white powder, crystals, Crystalline powder and/or chunks
- Infrared spectrum:Conforms to structure
- Titration:94.0%-106.0% (with morpholine)
- Gas liquid:94.0% (minimum)
Specification Test Results
About this item
:
:204.2219 g/mol
Melting Pt:70...75 °C (lit.)
EINECS:251-165-2
:GHS07
Storage Temperature:Room Temperature
CAS Number:32703-79-0
Formula:C₁₂H₁₂O₃
MDL Number:MFCD00060127