Tetrabutylammonium hydroxide 10% in water
Supplier: TCI
Certificates
About this item
CAS Number:2052-49-5
:
Formula:C₁₆H₃₇NO
MDL Number:MFCD00009425
:259.48 g/mol
Storage Temperature:Refrigerator
EINECS:218-147-6
UN:3267
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Neutralization titration):10.0 to 11.0 w/w%
- Ignition residue (Sulfate):max. 0.5 %
- Bromide:max. 0.5 %