Order Entry
Finland
ContactUsLinkComponent
Bromomethyl methyl ether tech. 90%, Technical Grade
Flammable
Health
Irritant
Bromomethyl methyl ether tech. 90%, Technical Grade
Catalog # L09837.06
CAS Number:  
Flammable
Health
Irritant
Bromomethyl methyl ether tech. 90%, Technical Grade
Catalog # L09837.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :124.97 g/mol
Boiling Pt:86…87 °C
Density:1.531
Flash Pt:26 °C (78 °F)
Storage Temperature:Refrigerator
UN:1992
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
CAS Number:13057-17-5
Formula:C₂H₅BrO
MDL Number:MFCD00000171