Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Titanium aluminide
Titanium aluminide ≥99.5% (metals basis) -325 mesh,
Titanium aluminide ≥99.5% (metals basis) -325 mesh,
:
Thermo Fisher Scientific
Titanium aluminide ≥99.5% (metals basis) -325 mesh,
:
Thermo Fisher Scientific
:
12003-96-2
:
:
AlTi
:
MFCD00210656
:
Ambient
:
3089
:
4.1,II
:
GHS02
:
GHS07
msds_ALFA088393
Thermo Scientific Acroseal 25 Years