
4-(Trifluoromethoxy)phenylhydrazine hydrochloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:133115-72-7
:
Formula:C₇H₈ClF₃N₂O
MDL Number:MFCD00053033
:228.60 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Potassium iodate Method):min. 98.0 %