
4-(Trifluoromethoxy)phenylhydrazine hydrochloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:133115-72-7
:
Formula:C₇H₈ClF₃N₂O
MDL Number:MFCD00053033
:228.60 g/mol
Melting Pt:∼ 225 °C (dec.)
EINECS:None
:GHS07
Storage Temperature:Ambient