0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
2-Acrylamido-2-methylpropanesulphonic acid
2-Acrylamido-2-methylpropanesulphonic acid ≥98%
2-Acrylamido-2-methylpropanesulphonic acid ≥98%
:
Thermo Fisher Scientific
2-Acrylamido-2-methylpropanesulphonic acid ≥98%
:
Thermo Fisher Scientific
:
15214-89-8
:
:
C₇H₁₃NO₄S
:
MFCD00007522
:
207.25 g/mol
:
∼ 195 °C (dec.)
:
1.10
:
239-268-0
:
8,III
:
GHS06
:
GHS05
msds_ALFAA18924
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years