Order Entry
Finland
ContactUsLinkComponent
HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥98.0% (by HPLC) coupling agent for peptides synthesis
HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥98.0% (by HPLC) coupling agent for peptides synthesis
Supplier:  TCI
Certificates
undefined
HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥98.0% (by HPLC) coupling agent for peptides synthesis
Supplier:  TCI
Certificates

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%

Specification Test Results