Order Entry
Finland
ContactUsLinkComponent
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
Catalog # MOLEM53412202
Supplier:  Molekula
CAS Number:  
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
Catalog # MOLEM53412202
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :380.2 g/mol
CAS Number:148893-10-1
Formula:C₁₀H₁₅F₆N₆OP
MDL Number:MFCD00274639