
O-Methylisourea hemisulphate ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:52328-05-9
:
Formula:C₄H₁₄N₄O₆S
MDL Number:MFCD00040594
:246.24 g/mol
EINECS:257-851-8
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (with Total Nitrogen):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together