Order Entry
Finland
ContactUsLinkComponent
 
 
 
3,5-Dimethylpiperidin (mixture of cis- and trans- isomers) cis- and trans-mixture ≥98.0% (by titrimetric analysis)
 :  TCI
 
undefined
 
 
3,5-Dimethylpiperidin (mixture of cis- and trans- isomers) cis- and trans-mixture ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 96.0 % (total of isomer)
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

Frequently Bought Together