Order Entry
Finland
ContactUsLinkComponent
Milk powder collector
Milk powder collector
Catalog # GERB3125
Supplier:  GERBER FUNKE
undefined
Milk powder collector
Catalog # GERB3125
Supplier:  GERBER FUNKE

Specifications

  • Ø:
    28 mm
  • Volume:
    230 ml
  • Description:
    Milk powder collector
  • Length:
    375 mm
  • Pk:
    1

Specifications

About this item

Stainless steel powder collector.

Specially designed for applications in milk industries.