Order Entry
Finland
ContactUsLinkComponent
Hydrometer for condensed milk
Hydrometer for condensed milk
Catalog # 614-0254
Supplier:  GERBER FUNKE
undefined
Hydrometer for condensed milk
Catalog # 614-0254
Supplier:  GERBER FUNKE

Specifications

  • Description:
    Hydrometer for condensed milk
  • Length:
    230 mm
  • Division:
    0,001 g/ml
  • Pk:
    1
  • Range:
    1,040 - 1,080 g/ml

Specifications

About this item

Hydrometer for density analysis of condensed milk.