Order Entry
Finland
ContactUsLinkComponent
Hoffmann clips, heavy duty
Hoffmann clips, heavy duty
Catalog # MURRL142/02
Supplier:  MURRAY S
Hoffmann clips, heavy duty
Catalog # MURRL142/02
Supplier:  MURRAY S
Specifications
  • Spanning width:
    40 mm
  • Pk:
    1
Specifications
About this item
Heavy duty clip fitted with nickel plated brass tommy screw and hinged bottom bar.
  • Spanning width = Width of gate