Order Entry
Finland
ContactUsLinkComponent
Sleeves for vacuum filtration
Sleeves for vacuum filtration
Catalog # KART420
Supplier:  KARTELL
undefined
Sleeves for vacuum filtration
Catalog # KART420
Supplier:  KARTELL
Specifications
  • Description:
    Set of 7, int. Ø 17 to 75 mm, ext. Ø 21 to 86 mm
  • Pk:
    1
Specifications
About this item
Set of 7 adapters, conical, made of black neoprene, sleeve supports for Büchner funnels and filter flasks.