HPLC columns, NUCLEOSIL® C18 HD
NUCLEOSIL® C18 HD is an octadecyl modified silica phase with a high density coverage, using for HPLC.
- Robust spherical silica gel
- Strongly hydrophobic, endcapped phase for RP chromatography
- Separation mechanism based on hydrophobic interactions
EC guard columns are used for analytical EC columns with the column protection system guard column holder (554-5851).
- Catalog No:
- MANA720191.40
- MANA720191.46
- MANA720296.46
- MANA721072.30
- MANA720294.46
- MANA720280.46
- MANA720193.46
- MANA720280.40
- MANA721196.30
- MANA720296.40
- Carbon load:
- 20%
- 20%
- 20%
- 20%
- 20%
- 20%
- 20%
- 20%
- 20%
- 20%
- Endcapped:
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- Yes, Yes
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- USP code:
- L1, L1
- L1, L1
- L1, L1
- L1, L1
- L1, L1
- L1, L1
- L1, L1
- L1, L1
- L1, L1
- L1, L1