Orders LinkContactUsLinkComponent
chromatography-46643-1920x760.jpg
Understanding analyte physicochemical properties and their value in LC method development
Webinar
Thursday, May 12, 2022
3:00 PM - 4:00 PM EDT (USA, Canada)
Replay
Understanding analyte physicochemical properties and their value in LC method development | Avantor
Email_Icon_RGB.png
Email alerts
Registration_Icon_RGB.png
Free registration
Chat_Icon_RGB.png
Chat area
Certified_Icon_RGB.png
Certificate of attendance
Analyte retention in liquid chromatography (LC) is driven by molecular interactions between analyte molecules and both the mobile phase and stationary phase.
The type and extent of these interactions are dependent on the physicochemical properties of the analyte. An understanding of analyte properties is invaluable for method development, enabling selection of the appropriate chromatographic mode and key method parameters (e.g. mobile phase and stationary phase selection). For example, analyte pKa, logP and logD values can be used to evaluate ionisation state and hydrophobicity and can help predict analyte retention and guide mobile phase composition, pH and buffer selection. This webinar will review the range of analyte properties that are important to consider when developing methods. These will be discussed in the context of the interactions between the analyte, mobile phase and stationary phase, which defines the separation process. Additionally, analyte structure presents opportunities to optimise separation selectivity by exploiting additional retention mechanisms offered by novel stationary phase chemistries. This will be discussed with examples demonstrating the impact this can have on the chromatographic separation obtained.
For more information,please contact us at [email protected]
Presented by
Tony-Edge100.jpg
Tony Edge
Tony Edge is R&D Manager at Avantor Sciences, heading a team of specialist scientists in manufacturing and developing next generation stationary phases for HPLC. He has worked in both manufacturing and also industry, having periods of employment at LGC and also AstraZeneca as well as ThermoFisherScientific and latterly Agilent Technologies. In 2008, he was fortunate enough to be awarded the Desty memorial lecture for his contributions to innovating separation science, and in the same year also won a clinical excellence award from AstraZeneca. He is currently the President of the UK chromatographic Society and also the chair of the permenent scientific committee for the ISC, as well as being the chair for the next ISC which is due to be held in Liverpool in 2024.