Orders LinkContactUsLinkComponent
chromatography-cs-1920x760.jpg
A UHPLC/HPLC Method Development Strategy with Complementary Stationary Phases to Maximise Selectivity and Resolution
Friday, July 12, 2019
16:00 to 16:45 CEST (Berlin, Paris, Madrid)
Register & watch
A UHPLC/HPLC Method Development Strategy with Complementary Stationary Phases to Maximize Selectivity and Resolution | Avantor
Email_Icon_RGB.png
Email alerts
Registration_Icon_RGB.png
Free registration
Chat_Icon_RGB.png
Chat area
Certified_Icon_RGB.png
Certificate of attendance
In this seminar, we review the importance of chromatographic selectivity in RPLC from a theoretical and practical perspective and how this relates to analyte resolution for method development. With an understanding of selectivity, and using a variety of chromatographic data, we discuss phase design principles and how it is possible to introduce functionality to enhance selectivity through mechanisms such as hydrophobicity, π-π, dipole-dipole and shape selectivity interactions.
An overview of the method development workflow is discussed. Based upon the key parameters to maximize selectivity, a systematic and optimized method development screening platform is described and an example related substances method development activity is illustrated using the complementary stationary phases with MeOH and MeCN solvents.
Presented by
Alan-McKeow-100.jpg
Alan McKeown
As an experienced chromatographer in his own right, Alan has been invited and delivered chromatography technical seminars in Europe, Asia, North America and South America at conferences, customer sites and organised events. Alan has published various peer-reviewed articles and written different book chapters on separation science, chromatography and genotoxic impurities.
Alan is a Fellow of the Royal Society of Chemistry and a Chartered Chemist.