Orders LinkContactUsLinkComponent
Novel-approaches-hero.jpg
Hydrophobic interaction chromatography: a versatile method for ADC and protein analysis
Webinar
Tuesday, 5th December, 2023  
15:00 to 16:00 CET
Replay
Hydrophobic interaction chromatography: a versatile method for ADC and protein analysis | Avantor
WebinarEmailAlerts.jpg
Email alerts
WebinarFreeRegistration.jpg
Free registration
Webinar-chat-area.jpg
Chat area
webinar-Certificate-of-attendance.jpg
Certificate of attendance
Hydrophobic interaction chromatography (HIC) is a powerful method for analyzing and characterizing protein-based therapeutics under native conditions. It is the gold standard method for antibody-drug-conjugate (ADCs) analyses, but is equally suitable for the analysis of protein modifications such as oxidations, post-translational modifications or deamidations in protein therapeutics such as monoclonal antibodies (mAbs). This webinar will explain how HIC works, present application examples, and provide tips and tricks for method development.
For more information, please contact us at [email protected]
Presented By:
ak-100.jpg
Andrea Krumm
Andrea Krumm studied biotechnology before gaining a PhD in cell- and cancer-biology. After four years as an applications specialist for optical analytical devices used in academic and pharmaceutical research, she joined Tosoh Bioscience GmbH in 2020 as a product manager for analytical columns. As such she is responsible for gathering customer requirements, application areas and addressing these with new and existing products of the analytical columns line and Tosoh’s Multi-Angle-Light-Scattering Detector LenS3.