Orders LinkContactUsLinkComponent
protection.png
How to implement an effective eye and face protection programme
Webinar
Thursday, 18th March, 2021
11:00 to 12:00 CET (Berlin, Paris, Madrid)
Replay
How to implement an effective eye and face protection programme | Avantor
WebinarEmailAlerts.jpg
Email alerts
WebinarFreeRegistration.jpg
Free registration
Webinar-chat-area.jpg
Chat area
webinar-Certificate-of-attendance.jpg
Certificate of attendance
Around 135 000 eye injuries in the workplace are reported every year in Europe, and many more go unreported. Studies show that 90% of eye injuries are preventable. In this webinar we will discuss the key elements of a successful eye and face protection programme: Understanding the hazards, matching the hazards with the protective features of eyewear or face shields, ensuring compatibility with other PPE, and providing effective user training.
The most important features of safety eyewear and face shields are the protective features as described by the EN 166 Standard. A secure fit and comfort are the next most important features as they are directly related to safety and compliance. In this webinar we will discuss the science behind the development of safety eyewear, and how 3M technologies aim for a comfortable and secure fit on a wide range of head shapes and sizes and for compatibility with other PPE.
For more information, please contact us at [email protected]
Presented by
Max.png
Max Holderbusch
Max Holderbusch is currently an EMEA Application Engineer in the R&D department of 3M's Personal Safety Division. An organic chemist by training, in his 32 year career with 3M, Max has helped customers from multiple industry segments solve their problems with 3M product solutions ranging from Pressure Sensitive Adhesives, Industrial Abrasives, Specialty Chemicals and Personal Protective Equipment. As a member of 3M's R&D community, Max has been intimately involved in the development of new products and technology, the last 5 years focused on new Head, Eye and Face protective equipment.